| M13mp19
Sequence M13MP19 See Ebright et. al. (1992)Gene 114,
81-83. They report seven differences to the original sequence published by Yanisch-Perron.
Specifically, there should be a T at 1663, C at 1737, G at 2220, T at 2224, T at 2225, T
at 2229 and G at 2710. No corrections in this sequence.
AATGCTACTA CTATTAGTAG AATTGATGCC ACCTTTTCAG
CTCGCGCCCC AAATGAAAAT 60
ATAGCTAAAC AGGTTATTGA CCATTTGCGA AATGTATCTA ATGGTCAAAC TAAATCTACT 120
CGTTCGCAGA ATTGGGAATC AACTGTTACA TGGAATGAAA CTTCCAGACA CCGTACTTTA 180
GTTGCATATT TAAAACATGT TGAGCTACAG CACCAGATTC AGCAATTAAG CTCTAAGCCA 240
TCCGCAAAAA TGACCTCTTA TCAAAAGGAG CAATTAAAGG TACTCTCTAA TCCTGACCTG 300
TTGGAGTTTG CTTCCGGTCT GGTTCGCTTT GAAGCTCGAA TTAAAACGCG ATATTTGAAG 360
TCTTTCGGGC TTCCTCTTAA TCTTTTTGAT GCAATCCGCT TTGCTTCTGA CTATAATAGT 420
CAGGGTAAAG ACCTGATTTT TGATTTATGG TCATTCTCGT TTTCTGAACT GTTTAAAGCA 480
TTTGAGGGGG ATTCAATGAA TATTTATGAC GATTCCGCAG TATTGGACGC TATCCAGTCT 540
AAACATTTTA CTATTACCCC CTCTGGCAAA ACTTCTTTTG CAAAAGCCTC TCGCTATTTT 600
GGTTTTTATC GTCGTCTGGT AAACGAGGGT TATGATAGTG TTGCTCTTAC TATGCCTCGT 660
AATTCCTTTT GGCGTTATGT ATCTGCATTA GTTGAATGTG GTATTCCTAA ATCTCAACTG 720
ATGAATCTTT CTACCTGTAA TAATGTTGTT CCGTTAGTTC GTTTTATTAA CGTAGATTTT 780
TCTTCCCAAC GTCCTGACTG GTATAATGAG CCAGTTCTTA AAATCGCATA AGGTAATTCA 840
CAATGATTAA AGTTGAAATT AAACCATCTC AAGCCCAATT TACTACTCGT TCTGGTGTTT 900
CTCGTCAGGG CAAGCCTTAT TCACTGAATG AGCAGCTTTG TTACGTTGAT TTGGGTAATG 960
AATATCCGGT TCTTGTCAAG ATTACTCTTG ATGAAGGTCA GCCAGCCTAT GCGCCTGGTC 1020
TGTACACCGT TCATCTGTCC TCTTTCAAAG TTGGTCAGTT CGGTTCCCTT ATGATTGACC 1080
GTCTGCGCCT CGTTCCGGCT AAGTAACATG GAGCAGGTCG CGGATTTCGA CACAATTTAT 1140
CAGGCGATGA TACAAATCTC CGTTGTACTT TGTTTCGCGC TTGGTATAAT CGCTGGGGGT 1200
CAAAGATGAG TGTTTTAGTG TATTCTTTCG CCTCTTTCGT TTTAGGTTGG TGCCTTCGTA 1260
GTGGCATTAC GTATTTTACC CGTTTAATGG AAACTTCCTC ATGAAAAAGT CTTTAGTCCT 1320
CAAAGCCTCT GTAGCCGTTG CTACCCTCGT TCCGATGCTG TCTTTCGCTG CTGAGGGTGA 1380
CGATCCCGCA AAAGCGGCCT TTAACTCCCT GCAAGCCTCA GCGACCGAAT ATATCGGTTA 1440
TGCGTGGGCG ATGGTTGTTG TCATTGTCGG CGCAACTATC GGTATCAAGC TGTTTAAGAA 1500
ATTCACCTCG AAAGCAAGCT GATAAACCGA TACAATTAAA GGCTCCTTTT GGAGCCTTTT 1560
TTTTTGGAGA TTTTCAACGT GAAAAAATTA TTATTCGCAA TTCCTTTAGT TGTTCCTTTC 1620
TATTCTCACT CCGCTGAAAC TGTTGAAAGT TGTTTAGCAA AACCCCATAC AGAAAATTCA 1680
TTTACTAACG TCTGGAAAGA CGACAAAACT TTAGATCGTT ACGCTAACTA TGAGGGTTGT 1740
CTGTGGAATG CTACAGGCGT TGTAGTTTGT ACTGGTGACG AAACTCAGTG TTACGGTACA 1800
TGGGTTCCTA TTGGGCTTGC TATCCCTGAA AATGAGGGTG GTGGCTCTGA GGGTGGCGGT 1860
TCTGAGGGTG GCGGTTCTGA GGGTGGCGGT ACTAAACCTC CTGAGTACGG TGATACACCT 1920
ATTCCGGGCT ATACTTATAT CAACCCTCTC GACGGCACTT ATCCGCCTGG TACTGAGCAA 1980
AACCCCGCTA ATCCTAATCC TTCTCTTGAG GAGTCTCAGC CTCTTAATAC TTTCATGTTT 2040
CAGAATAATA GGTTCCGAAA TAGGCAGGGG GCATTAACTG TTTATACGGG CACTGTTACT 2100
CAAGGCACTG ACCCCGTTAA AACTTATTAC CAGTACACTC CTGTATCATC AAAAGCCATG 2160
TATGACGCTT ACTGGAACGG TAAATTCAGA GACTGCGCTT TCCATTCTGG CTTTAATGAA 2220
GATCCATTCG TTTGTGAATA TCAAGGCCAA TCGTCTGACC TGCCTCAACC TCCTGTCAAT 2280
GCTGGCGGCG GCTCTGGTGG TGGTTCTGGT GGCGGCTCTG AGGGTGGTGG CTCTGAGGGT 2340
GGCGGTTCTG AGGGTGGCGG CTCTGAGGGA GGCGGTTCCG GTGGTGGCTC TGGTTCCGGT 2400
GATTTTGATT ATGAAAAGAT GGCAAACGCT AATAAGGGGG CTATGACCGA AAATGCCGAT 2460
GAAAACGCGC TACAGTCTGA CGCTAAAGGC AAACTTGATT CTGTCGCTAC TGATTACGGT 2520
GCTGCTATCG ATGGTTTCAT TGGTGACGTT TCCGGCCTTG CTAATGGTAA TGGTGCTACT 2580
GGTGATTTTG CTGGCTCTAA TTCCCAAATG GCTCAAGTCG GTGACGGTGA TAATTCACCT 2640
TTAATGAATA ATTTCCGTCA ATATTTACCT TCCCTCCCTC AATCGGTTGA ATGTCGCCCT 2700
TTTGTCTTTA GCGCTGGTAA ACCATATGAA TTTTCTATTG ATTGTGACAA AATAAACTTA 2760
TTCCGTGGTG TCTTTGCGTT TCTTTTATAT GTTGCCACCT TTATGTATGT ATTTTCTACG 2820
TTTGCTAACA TACTGCGTAA TAAGGAGTCT TAATCATGCC AGTTCTTTTG GGTATTCCGT 2880
TATTATTGCG TTTCCTCGGT TTCCTTCTGG TAACTTTGTT CGGCTATCTG CTTACTTTTC 2940
TTAAAAAGGG CTTCGGTAAG ATAGCTATTG CTATTTCATT GTTTCTTGCT CTTATTATTG 3000
GGCTTAACTC AATTCTTGTG GGTTATCTCT CTGATATTAG CGCTCAATTA CCCTCTGACT 3060
TTGTTCAGGG TGTTCAGTTA ATTCTCCCGT CTAATGCGCT TCCCTGTTTT TATGTTATTC 3120
TCTCTGTAAA GGCTGCTATT TTCATTTTTG ACGTTAAACA AAAAATCGTT TCTTATTTGG 3180
ATTGGGATAA ATAATATGGC TGTTTATTTT GTAACTGGCA AATTAGGCTC TGGAAAGACG 3240
CTCGTTAGCG TTGGTAAGAT TCAGGATAAA ATTGTAGCTG GGTGCAAAAT AGCAACTAAT 3300
CTTGATTTAA GGCTTCAAAA CCTCCCGCAA GTCGGGAGGT TCGCTAAAAC GCCTCGCGTT 3360
CTTAGAATAC CGGATAAGCC TTCTATATCT GATTTGCTTG CTATTGGGCG CGGTAATGAT 3420
TCCTACGATG AAAATAAAAA CGGCTTGCTT GTTCTCGATG AGTGCGGTAC TTGGTTTAAT 3480
ACCCGTTCTT GGAATGATAA GGAAAGACAG CCGATTATTG ATTGGTTTCT ACATGCTCGT 3540
AAATTAGGAT GGGATATTAT TTTTCTTGTT CAGGACTTAT CTATTGTTGA TAAACAGGCG 3600
CGTTCTGCAT TAGCTGAACA TGTTGTTTAT TGTCGTCGTC TGGACAGAAT TACTTTACCT 3660
TTTGTCGGTA CTTTATATTC TCTTATTACT GGCTCGAAAA TGCCTCTGCC TAAATTACAT 3720
GTTGGCGTTG TTAAATATGG CGATTCTCAA TTAAGCCCTA CTGTTGAGCG TTGGCTTTAT 3780
ACTGGTAAGA ATTTGTATAA CGCATATGAT ACTAAACAGG CTTTTTCTAG TAATTATGAT 3840
TCCGGTGTTT ATTCTTATTT AACGCCTTAT TTATCACACG GTCGGTATTT CAAACCATTA 3900
AATTTAGGTC AGAAGATGAA ATTAACTAAA ATATATTTGA AAAAGTTTTC TCGCGTTCTT 3960
TGTCTTGCGA TTGGATTTGC ATCAGCATTT ACATATAGTT ATATAACCCA ACCTAAGCCG 4020
GAGGTTAAAA AGGTAGTCTC TCAGACCTAT GATTTTGATA AATTCACTAT TGACTCTTCT 4080
CAGCGTCTTA ATCTAAGCTA TCGCTATGTT TTCAAGGATT CTAAGGGAAA ATTAATTAAT 4140
AGCGACGATT TACAGAAGCA AGGTTATTCA CTCACATATA TTGATTTATG TACTGTTTCC 4200
ATTAAAAAAG GTAATTCAAA TGAAATTGTT AAATGTAATT AATTTTGTTT TCTTGATGTT 4260
TGTTTCATCA TCTTCTTTTG CTCAGGTAAT TGAAATGAAT AATTCGCCTC TGCGCGATTT 4320
TGTAACTTGG TATTCAAAGC AATCAGGCGA ATCCGTTATT GTTTCTCCCG ATGTAAAAGG 4380
TACTGTTACT GTATATTCAT CTGACGTTAA ACCTGAAAAT CTACGCAATT TCTTTATTTC 4440
TGTTTTACGT GCTAATAATT TTGATATGGT TGGTTCAATT CCTTCCATAA TTCAGAAGTA 4500
TAATCCAAAC AATCAGGATT ATATTGATGA ATTGCCATCA TCTGATAATC AGGAATATGA 4560
TGATAATTCC GCTCCTTCTG GTGGTTTCTT TGTTCCGCAA AATGATAATG TTACTCAAAC 4620
TTTTAAAATT AATAACGTTC GGGCAAAGGA TTTAATACGA GTTGTCGAAT TGTTTGTAAA 4680
GTCTAATACT TCTAAATCCT CAAATGTATT ATCTATTGAC GGCTCTAATC TATTAGTTGT 4740
TAGTGCACCT AAAGATATTT TAGATAACCT TCCTCAATTC CTTTCTACTG TTGATTTGCC 4800
AACTGACCAG ATATTGATTG AGGGTTTGAT ATTTGAGGTT CAGCAAGGTG ATGCTTTAGA 4860
TTTTTCATTT GCTGCTGGCT CTCAGCGTGG CACTGTTGCA GGCGGTGTTA ATACTGACCG 4920
CCTCACCTCT GTTTTATCTT CTGCTGGTGG TTCGTTCGGT ATTTTTAATG GCGATGTTTT 4980
AGGGCTATCA GTTCGCGCAT TAAAGACTAA TAGCCATTCA AAAATATTGT CTGTGCCACG 5040
TATTCTTACG CTTTCAGGTC AGAAGGGTTC TATCTCTGTT GGCCAGAATG TCCCTTTTAT 5100
TACTGGTCGT GTGACTGGTG AATCTGCCAA TGTAAATAAT CCATTTCAGA CGATTGAGCG 5160
TCAAAATGTA GGTATTTCCA TGAGCGTTTT TCCTGTTGCA ATGGCTGGCG GTAATATTGT 5220
TCTGGATATT ACCAGCAAGG CCGATAGTTT GAGTTCTTCT ACTCAGGCAA GTGATGTTAT 5280
TACTAATCAA AGAAGTATTG CTACAACGGT TAATTTGCGT GATGGACAGA CTCTTTTACT 5340
CGGTGGCCTC ACTGATTATA AAAACACTTC TCAAGATTCT GGCGTACCGT TCCTGTCTAA 5400
AATCCCTTTA ATCGGCCTCC TGTTTAGCTC CCGCTCTGAT TCCAACGAGG AAAGCACGTT 5460
ATACGTGCTC GTCAAAGCAA CCATAGTACG CGCCCTGTAG CGGCGCATTA AGCGCGGCGG 5520
GTGTGGTGGT TACGCGCAGC GTGACCGCTA CACTTGCCAG CGCCCTAGCG CCCGCTCCTT 5580
TCGCTTTCTT CCCTTCCTTT CTCGCCACGT TCGCCGGCTT TCCCCGTCAA GCTCTAAATC 5640
GGGGGCTCCC TTTAGGGTTC CGATTTAGTG CTTTACGGCA CCTCGACCCC AAAAAACTTG 5700
ATTTGGGTGA TGGTTCACGT AGTGGGCCAT CGCCCTGATA GACGGTTTTT CGCCCTTTGA 5760
CGTTGGAGTC CACGTTCTTT AATAGTGGAC TCTTGTTCCA AACTGGAACA ACACTCAACC 5820
CTATCTCGGG CTATTCTTTT GATTTATAAG GGATTTTGCC GATTTCGGAA CCACCATCAA 5880
ACAGGATTTT CGCCTGCTGG GGCAAACCAG CGTGGACCGC TTGCTGCAAC TCTCTCAGGG 5940
CCAGGCGGTG AAGGGCAATC AGCTGTTGCC CGTCTCGCTG GTGAAAAGAA AAACCACCCT 6000
GGCGCCCAAT ACGCAAACCG CCTCTCCCCG CGCGTTGGCC GATTCATTAA TGCAGCTGGC 6060
ACGACAGGTT TCCCGACTGG AAAGCGGGCA GTGAGCGCAA CGCAATTAAT GTGAGTTAGC 6120
TCACTCATTA GGCACCCCAG GCTTTACACT TTATGCTTCC GGCTCGTATG TTGTGTGGAA 6180
TTGTGAGCGG ATAACAATTT CACACAGGAA ACAGCTATGA CCATGATTAC GCCAAGCTTG 6240
CATGCCTGCA GGTCGACTCT AGAGGATCCC CGGGTACCGA GCTCGAATTC ACTGGCCGTC 6300
GTTTTACAAC GTCGTGACTG GGAAAACCCT GGCGTTACCC AACTTAATCG CCTTGCAGCA 6360
CATCCCCCTT TCGCCAGCTG GCGTAATAGC GAAGAGGCCC GCACCGATCG CCCTTCCCAA 6420
CAGTTGCGCA GCCTGAATGG CGAATGGCGC TTTGCCTGGT TTCCGGCACC AGAAGCGGTG 6480
CCGGAAAGCT GGCTGGAGTG CGATCTTCCT GAGGCCGATA CGGTCGTCGT CCCCTCAAAC 6540
TGGCAGATGC ACGGTTACGA TGCGCCCATC TACACCAACG TAACCTATCC CATTACGGTC 6600
AATCCGCCGT TTGTTCCCAC GGAGAATCCG ACGGGTTGTT ACTCGCTCAC ATTTAATGTT 6660
GATGAAAGCT GGCTACAGGA AGGCCAGACG CGAATTATTT TTGATGGCGT TCCTATTGGT 6720
TAAAAAATGA GCTGATTTAA CAAAAATTTA ACGCGAATTT TAACAAAATA TTAACGTTTA 6780
CAATTTAAAT ATTTGCTTAT ACAATCTTCC TGTTTTTGGG GCTTTTCTGA TTATCAACCG 6840
GGGTACATAT GATTGACATG CTAGTTTTAC GATTACCGTT CATCGATTCT CTTGTTTGCT 6900
CCAGACTCTC AGGCAATGAC CTGATAGCCT TTGTAGATCT CTCAAAAATA GCTACCCTCT 6960
CCGGCATTAA TTTATCAGCT AGAACGGTTG AATATCATAT TGATGGTGAT TTGACTGTCT 7020
CCGGCCTTTC TCACCCTTTT GAATCTTTAC CTACACATTA CTCAGGCATT GCATTTAAAA 7080
TATATGAGGG TTCTAAAAAT TTTTATCCTT GCGTTGAAAT AAAGGCTTCT CCCGCAAAAG 7140
TATTACAGGG TCATAATGTT TTTGGTACAA CCGATTTAGC TTTATGCTCT GAGGCTTTAT 7200
TGCTTAATTT TGCTAATTCT TTGCCTTGCC TGTATGATTT ATTGGATGTT
7250
|