General Description
DNA pNMT41-CAT
Entire molecule length: 6719 bp
Restriction Map
Enzyme |
# of cuts |
Positions |
AatI |
1 |
3384 |
AccI |
5 |
1863 1985 2228 2992 4725 |
AccIII |
1 |
1357 |
AciI |
34 |
20(c) 407 1339 1703 1797(c) 3233 3245 3254 3266 3276 3287 3333 3946 4105 4261 4734(c) 4922(c) 4969 5068(c) 5177(c) 5254(c) 5298 5419(c) 5465 5656(c) 5747(c) 6109 6118(c) 6253 6363(c) 6484(c) 6503(c) 6630(c) 6658(c) |
AcsI |
12 |
75 1136 1361 1811 1896 2005 2151 2549 2719 2933 3037 4643 |
AcyI |
1 |
5273 |
AflII |
3 |
128 3114 3740 |
AflIII |
3 |
307 3055 4162 |
AgeI |
1 |
4805 |
AluI |
21 |
963 1261 1390 2313 2373 2477 3349 3403 3510 4002 4584 4706 4891 5460 5523 5623 6144 6401 6447 6537 6705 |
Alw44I |
2 |
5141 6387 |
AlwI |
11 |
421 450 4106(c) 5174 5178(c) 5495 5958(c) 5959 6055(c) 6057 6143 |
AlwNI |
2 |
561 6292 |
AosI |
1 |
5590 |
ApaLI |
2 |
5141 6387 |
ApoI |
12 |
75 1136 1361 1811 1896 2005 2151 2549 2719 2933 3037 4643 |
AseI |
8 |
295 517 1092 1885 2773 3153 4791 5638 |
AsnI |
8 |
295 517 1092 1885 2773 3153 4791 5638 |
Asp700 |
2 |
4062 5213 |
AspEI |
2 |
1127 5813 |
AspHI |
3 |
5145 5230 6391 |
AsuII |
1 |
3547 |
AvaII |
5 |
1021 3660 3988 5449 5671 |
AviII |
1 |
5590 |
AvrII |
2 |
1949 3385 |
BanI |
1 |
5860 |
BbsI |
1 |
3877 |
BbvI |
10 |
1008 3557(c) 4334(c) 4571(c) 5379(c) 5770 6073(c) 6279(c) 6282(c) 6372 |
BcgI |
1 |
5275(c) |
BfaI |
12 |
779 1950 2314 2584 2997 3002 3386 4116 4547 5620 5955 6208 |
BfrI |
3 |
128 3114 3740 |
BglI |
1 |
5695 |
BlnI |
2 |
1949 3385 |
BmyI |
3 |
5145 5230 6391 |
BpmI |
4 |
441 1483 4282(c) 5744 |
Bpu1102I |
1 |
2478 |
BpuAI |
1 |
3877 |
BsaHI |
1 |
5273 |
BsaI |
2 |
1850 5747 |
BsaJI |
16 |
455 1592 1593 1662 1840 1949 2049 3292 3327 3385 3991 4202 4210 4234 4286 6541 |
BsaWI |
6 |
1011 1357 4805 5517 6348 6495 |
BseAI |
1 |
1357 |
BsgI |
1 |
1015 |
BsiEI |
3 |
5295 5444 6367 |
BsiHKAI |
3 |
5145 5230 6391 |
BsiYI |
13 |
718 1018 1593 1841 3293 3669 3696 4287 4386 6223 6502 6668 6686 |
BslI |
13 |
718 1018 1593 1841 3293 3669 3696 4287 4386 6223 6502 6668 6686 |
BsmAI |
8 |
736(c) 798 847(c) 1585 1850 4533 4971(c) 5747 |
BsmFI |
2 |
3215(c) 3576(c) |
BsmI |
4 |
1354 1761 2442 2850 |
Bsp1286I |
3 |
5145 5230 6391 |
BspEI |
1 |
1357 |
BspHI |
3 |
2837 4973 5981 |
BspMI |
1 |
4419(c) |
BspWI |
17 |
1139 1702 1935 2823 3253 3332 3338 3579 3615 3954 3977 4236 4543 4888 5695 6083 6655 |
BsrBI |
1 |
4971(c) |
BsrDI |
7 |
996 1380 2955 3975(c) 4357 5579 5753(c) |
BsrFI |
2 |
4805 5728 |
BsrGI |
1 |
2180 |
BsrI |
17 |
424 911 938 1168 1608(c) 3270(c) 3680 4445 4538(c) 5168 5338(c) 5607 5650 5768 6174 6286(c) 6299(c) |
Bst1107I |
2 |
1986 2993 |
BstBI |
1 |
3547 |
BstEII |
1 |
3459 |
BstNI |
8 |
1063 1538 1594 3457 4204 6542 6555 6676 |
BstUI |
6 |
2320 4922 5254 5747 6077 6658 |
BstXI |
2 |
589 4140 |
BstYI |
6 |
5166 5183 5951 5963 6049 6060 |
CelII |
1 |
2478 |
CfoI |
13 |
601 2320 2880 3611 4922 5254 5591 5684 6077 6186 6360 6460 6527 |
Cfr10I |
2 |
4805 5728 |
Csp45I |
1 |
3547 |
Csp6I |
18 |
702 1239 1777 1992 2181 2798 2871 2895 3119 3673 3731 3811 4274 4465 4541 4666 5331 6709 |
DdeI |
12 |
86 144 1582 2478 2874 3345 3436 5312 5852 6018 6427 6718 |
DpnI |
19 |
415 444 2509 3444 4113 4652 5132 5168 5185 5443 5489 5507 5848 5953 5965 6043 6051 6062 6137 |
DpnII |
19 |
413 442 2507 3442 4111 4650 5130 5166 5183 5441 5487 5505 5846 5951 5963 6041 6049 6060 6135 |
DraI |
9 |
614 1281 1620 3019 4802 4852 5235 5927 5946 |
DraII |
1 |
6714 |
DrdI |
1 |
6599 |
DsaI |
3 |
1662 3292 4286 |
DsaV |
11 |
1061 1536 1592 3455 4202 5275 5626 6322 6540 6553 6674 |
EaeI |
3 |
1626 3943 5420 |
Eam1105I |
2 |
1127 5813 |
EarI |
3 |
1047(c) 4022 5014(c) |
Eco47III |
1 |
2879 |
Eco57I |
4 |
4402 4877(c) 5147 6159(c) |
EcoNI |
2 |
3694 4384 |
EcoO109I |
1 |
6714 |
EcoRI |
3 |
1136 1361 1811 |
EcoRII |
8 |
1061 1536 1592 3455 4202 6540 6553 6674 |
Esp3I |
2 |
736(c) 1585 |
EspI |
1 |
2478 |
Fnu4HI |
19 |
997 1703 3333 3571 3946 4105 4348 4585 5069 5298 5393 5420 5759 6087 6293 6296 6361 6504 6659 |
FnuDII |
6 |
2320 4922 5254 5747 6077 6658 |
FokI |
10 |
820 922 1341(c) 3236(c) 3746(c) 4437 4871 5374 5661 5842 |
FspI |
1 |
5590 |
HaeII |
3 |
2881 3612 6461 |
HaeIII |
20 |
1274 1319 1541 1628 3326 3332 3384 3945 3957 4014 4239 4395 5422 5689 5769 6227 6661 6679 6690 6715 |
HgaI |
5 |
2309(c) 4219(c) 5281 6011(c) 6589(c) |
HgiAI |
3 |
5145 5230 6391 |
HhaI |
13 |
601 2320 2880 3611 4922 5254 5591 5684 6077 6186 6360 6460 6527 |
HinP1I |
13 |
599 2318 2878 3609 4920 5252 5589 5682 6075 6184 6358 6458 6525 |
HincII |
3 |
270 1864 4322 |
HindII |
3 |
270 1864 4322 |
HindIII |
2 |
961 3401 |
HinfI |
11 |
971 1058 1711 1850 2533 3765 3931 4098 4759 5814 6331 |
HpaII |
14 |
1012 1316 1358 1486 4806 5276 5518 5628 5695 5729 6133 6323 6349 6496 |
HphI |
21 |
635 912(c) 1397 1403(c) 1405 1541 1597(c) 1609 1651(c) 3061(c) 3471 3891 4145(c) 4182 4455 4491 5086(c) 5121 5327(c) 5743 5970 |
ItaI |
19 |
997 1703 3333 3571 3946 4105 4348 4585 5069 5298 5393 5420 5759 6087 6293 6296 6361 6504 6659 |
Ksp632I |
3 |
1047(c) 4022 5014(c) |
MaeI |
12 |
779 1950 2314 2584 2997 3002 3386 4116 4547 5620 5955 6208 |
MaeII |
13 |
82 337 745 891 1448 1623 2560 3466 4722 4774 5211 5584 6000 |
MaeIII |
16 |
892 918 1418 1523 2561 3177 3459 3750 5152 5340 5493 5551 5882 6165 6281 6344 |
MboI |
19 |
413 442 2507 3442 4111 4650 5130 5166 5183 5441 5487 5505 5846 5951 5963 6041 6049 6060 6135 |
MboII |
20 |
109 671 1054 1064 1636(c) 2534(c) 2853 2954 3452 3882 4009(c) 4395 4413 4506 5031 5140 5218 5973 6044(c) 6196(c) |
McrI |
3 |
5295 5444 6367 |
MfeI |
5 |
1075 2570 2748 3726 4089 |
MluNI |
1 |
1628 |
MnlI |
32 |
532 616 619(c) 654 680 1048(c) 1207(c) 1845 1850 2701(c) 2970 3316(c) 3322(c) 3346 3352 3359(c) 3362(c) 3374(c) 3599(c) 3629 3837(c) 4025 4037(c) 4202 4250 5440(c) 5646(c) 5793 5874 6274 6524(c) 6598 |
MroI |
1 |
1357 |
MscI |
1 |
1628 |
MseI |
56 |
64 79 113 129 282 295 316 509 517 613 877 965 1028 1092 1280 1619 1761 1885 2029 2085 2388 2429 2665 2676 2703 2734 2739 2773 2832 2906 3018 3115 3125 3153 3196 3496 3505 3558 3741 4120 4411 4501 4517 4755 4791 4801 4810 4851 5234 5599 5638 5873 5926 5940 5945 5997 |
MslI |
6 |
637 3939 4138 5042 5401 5560 |
MspA1I |
6 |
20 1261 1705 5177 6118 6363 |
MspI |
14 |
1012 1316 1358 1486 4806 5276 5518 5628 5695 5729 6133 6323 6349 6496 |
MunI |
5 |
1075 2570 2748 3726 4089 |
MvaI |
8 |
1063 1538 1594 3457 4204 6542 6555 6676 |
MvnI |
6 |
2320 4922 5254 5747 6077 6658 |
MwoI |
17 |
1139 1702 1935 2823 3253 3332 3338 3579 3615 3954 3977 4236 4543 4888 5695 6083 6655 |
NciI |
3 |
5277 5628 6324 |
NcoI |
2 |
1662 3292 |
NdeII |
19 |
413 442 2507 3442 4111 4650 5130 5166 5183 5441 5487 5505 5846 5951 5963 6041 6049 6060 6135 |
NlaIII |
27 |
152 311 636 721 1436 1666 1727 1748 2093 2227 2699 2841 3059 3296 3751 3956 3969 4112 4166 4285 4848 4977 5370 5406 5484 5494 5985 |
NlaIV |
9 |
4208 4457 4926 5516 5727 5768 5862 6634 6673 |
NsiI |
1 |
4864 |
NspI |
6 |
152 311 2093 3059 4166 4848 |
NspV |
1 |
3547 |
PflMI |
2 |
718 1593 |
PinAI |
1 |
4805 |
PleI |
3 |
3759(c) 5822 6325(c) |
Ppu10I |
1 |
4860 |
Psp1406I |
3 |
1448 5211 5584 |
PvuI |
1 |
5444 |
PvuII |
1 |
1261 |
RcaI |
3 |
2837 4973 5981 |
RsaI |
18 |
703 1240 1778 1993 2182 2799 2872 2896 3120 3674 3732 3812 4275 4466 4542 4667 5332 6710 |
SalI |
1 |
1862 |
Sau3AI |
19 |
413 442 2507 3442 4111 4650 5130 5166 5183 5441 5487 5505 5846 5951 5963 6041 6049 6060 6135 |
Sau96I |
9 |
1021 3660 3956 3988 5449 5671 5688 5767 6714 |
ScaI |
7 |
1778 1993 2799 2896 4542 4667 5332 |
ScrFI |
11 |
1063 1538 1594 3457 4204 5277 5628 6324 6542 6555 6676 |
SfaNI |
20 |
801(c) 1204 1689(c) 1916(c) 2364 2577(c) 2935(c) 3214 3569(c) 3623(c) 3768 4394(c) 4415(c) 4472(c) 4849(c) 4871 5112(c) 5361 5552(c) 6604(c) |
SfcI |
5 |
2072 3583 5567 6245 6436 |
SfuI |
1 |
3547 |
SnoI |
2 |
5141 6387 |
SspBI |
1 |
2180 |
SspI |
6 |
1673 2027 2852 3024 4701 5008 |
StuI |
1 |
3384 |
StyI |
9 |
455 1662 1949 2049 3292 3385 3991 4210 4234 |
TaqI |
11 |
340 683 1032 1848 1863 1924 3541 3547 4708 5159 6603 |
TfiI |
8 |
971 1058 1711 1850 2533 3931 4098 4759 |
ThaI |
6 |
2320 4922 5254 5747 6077 6658 |
Tru9I |
56 |
64 79 113 129 282 295 316 509 517 613 877 965 1028 1092 1280 1619 1761 1885 2029 2085 2388 2429 2665 2676 2703 2734 2739 2773 2832 2906 3018 3115 3125 3153 3196 3496 3505 3558 3741 4120 4411 4501 4517 4755 4791 4801 4810 4851 5234 5599 5638 5873 5926 5940 5945 5997 |
Tsp509I |
67 |
75 213 274 313 362 514 550 585 609 782 823 1075 1089 1093 1136 1361 1766 1811 1886 1896 1978 2005 2065 2078 2151 2278 2324 2365 2378 2389 2405 2442 2492 2549 2570 2596 2634 2672 2719 2727 2735 2748 2765 2774 2833 2907 2933 3037 3044 3090 3108 3154 3211 3303 3559 3726 3798 3834 4089 4362 4643 4792 4853 4884 5380 5635 5941 |
Van91I |
2 |
718 1593 |
XbaI |
3 |
778 2583 3001 |
XcmI |
1 |
3982 |
XhoII |
6 |
5166 5183 5951 5963 6049 6060 |
XmnI |
2 |
4062 5213 |
No cuts: AatII, Acc65I, ApaI, AscI, Asp718, AspI, AvaI, BamHI, BanII, BbrPI, BclI, BglII, BsaAI, BsaBI, BsiWI, Bsp120I, BspDI, BssHII, Bsu36I, ClaI, DraIII, EagI, Ecl136II, EclXI, EcoRV, HpaI, KasI, KpnI, KspI, MamI, MluI, NaeI, NarI, NdeI, NgoMI, NheI, NotI, NruI, PacI, PaeR7I, PmaCI, PmeI, PmlI, PpuMI, PstI, RsrII, SacI, SacII, SapI, SexAI, SfiI, SgrAI, SmaI, SnaBI, SpeI, SphI, SwaI, Tth111I, XhoI, XmaI, XmaIII