Plasmid pSPL3 from the New Exon Trapping System.
    It is designed to search genomic DNA for potential exon sequences. This sequence has not
    been confirmed by sequence analysis. It was assembled from the known sequence of fragments
    used to construct the vector.
    CTGTGGAATG TGTGTCAGTT AGGGTGTGGA AAGTCCCCAG
    GCTCCCCAGC AGGCAGAAGT   60
    ATGCAAAGCA TGCATCTCAA TTAGTCAGCA ACCAGGTGTG GAAAGTCCCC AGGCTCCCCA  120
    GCAGGCAGAA GTATGCAAAG CATGCATCTC AATTAGTCAG CAACCATAGT CCCGCCCCTA  180
    ACTCCGCCCA TCCCGCCCCT AACTCCGCCC AGTTCCGCCC ATTCTCCGCC CCATGGCTGA  240
    CTAATTTTTT TTATTTATGC AGAGGCCGAG GCCGCCTCGG CCTCTGAGCT ATTCCAGAAG  300
    TAGTGAGGAG GCTTTTTTGG AGGCCTAGGC TTTTGCAAAA AGCTTGGACT GTGTTTACTT  360
    GCAATCCCCC AAAACAGACA GAATGGTGCA TCTGTCCAGT GAGGAGAAGT CTGCGGTCAC  420
    TGCCCTGTGG GGCAAGGTGA ATGTGGAAGA AGTTGGTGGT GAGGCCCTGG GCAGGCTGCT  480
    GGTTGTCTAC CCATGGACCC AGAGGTTCTT CGAGTCCTTT GGGGACCTGT CCTCTGCAAA  540
    TGCTGTTATG AACAATCCTA AGGTGAAGGC TCATGGCAAG AAGGTGCTGG CTGCCTTCAG  600
    TGAGGGTCTG AGTCACCTGG ACAACCTCAA AGGCACCTTT GCTAAGCTGA GTGAACTGCA  660
    CTGTGACAAG CTGCACGTGC TCTAGAGTCG ACCCAGCAGT AAGTAATACA TGTAATGCAA  720
    CCTATACAAA TAGCAATAGT AGCATTAGTA GTAGCAATAA TAATAGCAAT AGTTGTGTGG  780
    TCCATAGTAA TCATAGAATA TAGGAAAATA TTAAGACAAA GAAAAATAGA CAGGTTAATT  840
    GATAGACTAA TAGAAAGAGC AGAAGACAGT GGCAATGAGA GTGAAGGAGA AATATCAGCA  900
    CTTGTGGAGA TGGGGGTGGA GATGGGGCAC CATGCTCCTT GGGATGTTGA TGATCTGTAG  960
    TGCTACAGAA AAATTGTGGG TCACAGTCTA TTATGGGGTA CGGGATCACC AGAATTCTGG 1020
    AGCTCGAGCG GCCGCTGCAG GATCCCAGAT ATCTGGTGAT CCCGTACCTG TGTGGAAGGA 1080
    AGCAACCACC ACTCTATTTT GTGCATCAGA TGCTAAAGCA TATGATACAG AGGTACATAA 1140
    TGTTTGGGCC ACACATGCCG GTGTACCCAC AGACCCCAAC CCACAAGAAG TAGTATTGGT 1200
    AAATGTGACA GAAAATTTTA ACATGTGGAA AAATGACATG GTAGAACAGA TGCATGAGGA 1260
    TATAATCAGT TTATGGGATC AAAGCCTAAA GCCATGTGTA AAATTAACCC CACTCTGTGT 1320
    TAGTTTAAAG TGCACTGATT TGAAGAATGA TACTAATACC AATAGTAGTA GCGGGAGAAT 1380
    GATAATGGAG AAAGGAGAGA TAAAAAACTG CTCTTTCAAT ATCAGCACAA GCATAAGAGG 1440
    TAAGGTGCAG AAAGAATATG CATTTTTTTA TAAACTTGAT ATAATACCAA TAGATAATGA 1500
    TACTACCAGC TATACGTTGA CAAGTTGTAA CACCTCAGTC ATTACACAGG CCTGTCCAAA 1560
    GGTATCCTTT GAGCCAATTC CCATACATTA TTGTGCCCCG GCTGGTTTTG CGATTCTAAA 1620
    ATGTAATAAT AAGACGTTCA ATGGAACAGG ACCATGTACA AATGTCAGCA CAGTACAATG 1680
    TACACATGGA ATTAGGCCAG TAGTATCAAC TCAACTGCTG TTAAATGGCA GTCTAGCAGA 1740
    AGAAGAGGTA GTAATTAGAT CTGTCAATTT CACGGACAAT GCTAAAACCA TAATAGTACA 1800
    GCTGAACACA TCTGTAGAAA TTAATTGTAC AAGACCCAAC AACAATACAA GAAAAAAAAT 1860
    CCGTATCCAG AGGGGACCAG GGAGAGCATT TGTTACAATA GGAAAAATAG GAAATATGAG 1920
    ACAAGCACAT TGTAACATTA GTAGAGCAAA ATGGAATGCC ACTTTAAAAC AGATAGCTAG 1980
    CAAATTAAGA GAACAATTTG GAAATAATAA AACAATAATC TTTAAGCAAT CCTCAGGAGG 2040
    GGACCCAGAA ATTGTAACGC ACAGTTTTAA TTGTGGAGGG GAATTTTTCT ACTGTAATTC 2100
    AACACAACTG TTTAATAGTA CTTGGTTTAA TAGTACTTGG AGTACTGAAG GGTCAAATAA 2160
    CACTGAAGGA AGTGACACAA TCACACTCCC ATGCAGAATA AAACAATTTA TAAACATGTG 2220
    GCAGGAAGTA GGAAAAGCAA TGTATGCCCC TCCCATCAGC GGACAAATTA GATGTTCATC 2280
    AAATATTACA GGGCTGCTAT TAACAAGAGA TGGTGGTAAT AACAACAATG GGTCCGAGAT 2340
    CTTCAGACCT GGAGGAGGAG ATATGAGGGA CAATTGGAGA AGTGAATTAT ATAAATATAA 2400
    AGTAGTAAAA ATTGAACCAT TAGGAGTAGC ACCCACCAAG GCAAAGAGAA GAGTGGTGCA 2460
    GAGAGAAAAA AGAGCAGTGG GAATAGGAGC TTTGTTCCTT GGGTTCTTGG GAGCAGCAGG 2520
    AAGCACTATG GGCGCAGCGT CAATGACGCT GACGGTACAG GCCAGACAAT TATTGTCTGG 2580
    TATAGTGCAG CAGCAGAACA ATTTGCTGAG GGCTATTGAG GCGCAACAGC ATCTGTTGCA 2640
    ACTCACAGTC TGGGGCATCA AGCAGCTCCA GGCAAGAATC CTGGCTGTGG AAAGATACCT 2700
    AAAGGATCAA CAGCTCCTGG GGATTTGGGG TTGCTCTGGA AAACTACTTT GCACCACTGC 2760
    TGTGCCTTGG AATGCTAGTT GGAGTAATAA ATCTCTGGAA CAGATTTGGA ATCACACGAC 2820
    GTGGATGGAG TGGGACAGAG AAATTAACAA TTACACAAGC TTAATACACT CCTTAATTGA 2880
    AGAATCGCAA AACCAGCAAG AAAAGAATGA ACAAGAATTA TTGGAATTAG ATAAATGGGC 2940
    AAGTTTGTGG AATTGGTTTA ACATAACAAA TTGGCTGTGG TATATAAAAT TATTCATAAT 3000
    GATAGTAGGA GGCTTGGTAG GTTTAAGAAT AGTTTTTGCT GTACTTTCTG TAGTGAATAG 3060
    AGTTAGGCAG GGATATTCAC CATTATCGTT TCAGACCTGG AGATCTCCCG AGGGGACCCG 3120
    ACAGGCCCGA AGGAATAGAA GAAGAAGGTG GAGAGAGAGA CAGAGACAGA TCCATTTCGA 3180
    CCAATTCACT CCTCAGGTGC AGGCTGCCTA TCAGAAGGTG GTGGCTGGTG TGGCCAATGC 3240
    CCTGGCTCAC AAATACCACT GAGATCCAGA CATGATAAGA TACATTGATG AGTTTGGACA 3300
    AACCACAACT AGAATGCAGT GAAAAAAATG CTTTATTTGT GAAATTTGTG ATGCTATTGC 3360
    TTTATTTGTA ACCATTATAA GCTGCAATAA ACAAGTTAAC AACAACAATT GCATTCATTT 3420
    TATGTTTCAG GTTCAGGGGG AGGTGTGGGA GGTTTTTTAA AGCAAGTAAA ACCTCTACAA 3480
    ATGTGGTATG GCTGATTATG ATCCCCAGGA AGCTCCTCTG TGTCCTCATA AACCCTAACC 3540
    TCCTCTACTT GAGAGGACAT TCCAATCATA GGCTGCCCAT CCACCCTCTG TGTCCTCCTG 3600
    TTAATTAGGT CACTTAACAA AAAGGAAATT GGGTAGGGGT TTTTCACAGA CCGCTTTCTA 3660
    AGGGTAATTT TAAAATATCT GGGAAGTCCC TTCCACTGCT GTGTTCCAGA AGTGTTGGTA 3720
    AACAGCCCAC AAATGTCAAC AGCAGAAACA TACAAGCTGT CAGCTTTGCA CAAGGGCCCA 3780
    ACACCCTGCT CATCAAGAAG CACTGTGGTT GCTGTGTTAG TAATGTGCAA AACAGGAGGC 3840
    ACATTTTCCC CACCTGTGTA GGTTCCAAAA TATCTAGTGT TTTCATTTTT ACTTGGATCA 3900
    GGAACCCAGC ACTCCACTGG ATAAGCATTA TCCTTATCCA AAACAGCCTT GTGGTCAGTG 3960
    TTCATCTGCT GACTGTCAAC TGTAGCATTT TTTGGGGTTA CAGTTTGAGC AGGATATTTG 4020
    GTCCTGTAGT TTGCTAACAC ACCCCAGGTG GCACTTTTCG GGGAAATGTG CGCGGAACCC 4080
    CTATTTGTTT ATTTTTCTAA ATACATTCAA ATATGTATCC GCTCATGAGA CAATAACCCT 4140
    GATAAATGCT TCAATAATAT TGAAAAAGGA AGAGTATGAG TATTCAACAT TTCCGTGTCG 4200
    CCCTTATTCC CTTTTTTGCG GCATTTTGCC TTCCTGTTTT TGCTCACCCA GAAACGCTGG 4260
    TGAAAGTAAA AGATGCTGAA GATCAGTTGG GTGCACGAGT GGGTTACATC GAACTGGATC 4320
    TCAACAGCGG TAAGATCCTT GAGAGTTTTC GCCCCGAAGA ACGTTTTCCA ATGATGAGCA 4380
    CTTTTAAAGT TCTGCTATGT GGCGCGGTAT TATCCCGTAT TGACGCCGGG CAAGAGCAAC 4440
    TCGGTCGCCG CATACACTAT TCTCAGAATG ACTTGGTTGA GTACTCACCA GTCACAGAAA 4500
    AGCATCTTAC GGATGGCATG ACAGTAAGAG AATTATGCAG TGCTGCCATA ACCATGAGTG 4560
    ATAACACTGC GGCCAACTTA CTTCTGACAA CGATCGGAGG ACCGAAGGAG CTAACCGCTT 4620
    TTTTGCACAA CATGGGGGAT CATGTAACTC GCCTTGATCG TTGGGAACCG GAGCTGAATG 4680
    AAGCCATACC AAACGACGAG CGTGACACCA CGATGCCTGT AGCAATGGCA ACAACGTTGC 4740
    GCAAACTATT AACTGGCGAA CTACTTACTC TAGCTTCCCG GCAACAATTA ATAGACTGGA 4800
    TGGAGGCGGA TAAAGTTGCA GGACCACTTC TGCGCTCGGC CCTTCCGGCT GGCTGGTTTA 4860
    TTGCTGATAA ATCTGGAGCC GGTGAGCGTG GGTCTCGCGG TATCATTGCA GCACTGGGGC 4920
    CAGATGGTAA GCCCTCCCGT ATCGTAGTTA TCTACACGAC GGGGAGTCAG GCAACTATGG 4980
    ATGAACGAAA TAGACAGATC GCTGAGATAG GTGCCTCACT GATTAAGCAT TGGTAACTGT 5040
    CAGACCAAGT TTACTCATAT ATACTTTAGA TTGATTTAAA ACTTCATTTT TAATTTAAAA 5100
    GGATCTAGGT GAAGATCCTT TTTGATAATC TCATGACCAA AATCCTTAAC GGTGAGTTTT 5160
    CGTTCCACTG AGCGTCAGAC CCCGTAGAAA AGATCAAAGG ATCTTCTTGA GATCCTTTTT 5220
    TTCTGCGCGT AATCTGCTGC TTGCAAACAA AAAAACCACC GCTACCAGCG GTGGTTTGTT 5280
    TGCCGGATCA AGAGCTACCA ACTCTTTTTC CGAAGGTAAC TGGCTTCAGC AGAGCGCAGA 5340
    TACCAAATAC TGTCCTTCTA GTGTAGCCGT AGTTAGGCCA CCACTTCAAG AACTCTGTAG 5400
    CACCGCCTAC ATACCTCGCT CTGCTAATCC TGTTACCAGT GGCTGCTGCC AGTGGCGATA 5460
    AGTCGTGTCT TACCGGGTTG GACTCAAGAC GATAGTTACC GGATAAGGCG CAGCGGTCGG 5520
    GCTGAACGGG GGGTTCGTGC ACACAGCCCA GCTTGGAGCG AACGACCTAC ACCGAACTGA 5580
    GATACCTACA GCGCGAGCAT TGAGAAAGCG CCACGCTTCC CGAAGGGAGA AAGGCGGACA 5640
    GGTATCCGGT AAGCGGCAGG GTCGGAACAG GAGAGCGCAC GAGGGAGCTT CCAGGGGGAA 5700
    ACGCCTGGTA TCTTTATAGT CCTGTCGGGT TTCGCCACCT CTGACTTGAG CGTCGATTTT 5760
    TGTGATGCTC GTCAGGGGGG CGGAGCCTAT GGAAAAACGC CAGCAACGCG GCCTTTTTAC 5820
    GGTTCCTGGC CTTTTGCTGG CCTTTTGCTC ACATGTTCTT TCCTGCGTTA TCCCCTGATT 5880
    CTGTGGATAA CCGTATTACC GCCTTTGAGT GAGCTGATAC CGCTCGCCGC AGCCGAACGA 5940
    CCGAGCGCAG CGAGTCAGTG AGCGAGGAAG CGGAAGAGCG CCCAATACGC AAACCGCCTC 6000
    TCCCCGCGCG TTGGCCGATT CATTAATGCA G
    6031