| pPROEX HT-Cat
Sequence This sequence has not been confirmed by
sequence analysis. It was assembled from the known sequence of fragments used to construct
the vector.
pPROEX HT-Cat,
control from the pPROEX HT Prokaryotic Expression System
Last Changed 10 DEC-1996
GTTTGACAGC TTATCATCGA CTGCACGGTG CACCAATGCT TCTGGCGTCA GGCAGCCATC 60
GGAAGCTGTG GTATGGCTGT GCAGGTCGTA AATCACTGCA TAATTCGTGT CGCTCAAGGC 120
GCACTCCCGT TCTGGATAAT GTTTTTTGCG CCGACATCAT AACGGTTCTG GCAAATATTC 180
TGAAATGAGC TGTTGACAAT TAATCATCCG GTCCGTATAA TCTGTGGAAT TGTGAGCGGA 240
TAACAATTTC ACACAGGAAA CAGACCATGT CGTACTACCA TCACCATCAC CATCACGATT 300
ACGATATCCC AACGACCGAA AACCTGTATT TTCAGGGCGG GGATCCAATG GAGAAAAAAA 360
TCACTGGATA TACCACCGTT GATATATCCC AATGGCATCG TAAAGAACAT TTTGAGGCAT 420
TTCAGTCAGT TGCTCAATGT ACCTATAACC AGACCGTTCA GCTGGATATT ACGGCCTTTT 480
TAAAGACCGT AAAGAAAAAT AAGCACAAGT TTTATCCGGC CTTTATTCAC ATTCTTGCCC 540
GCCTGATGAA TGCTCATCCG GAATTCCGTA TGGCAATGAA AGACGGTGAG CTGGTGATAT 600
GGGATAGTGT TCACCCTTGT TACACCGTTT TCCATGAGCA AACTGAAACG TTTTCATCGC 660
TCTGGAGTGA ATACCACGAC GATTTCCGGC AGTTTCTACA CATATATTCG CAAGATGTGG 720
CGTGTTACGG TGAAAACCTG GCCTATTTCC CTAAAGGGTT TATTGAGAAT ATGTTTTTCG 780
TCTCAGCCAA TCCCTGGGTG AGTTTCACCA GTTTTGATTT AAACGTGGCC AATATGGACA 840
ACTTCTTCGC CCCCGTTTTC ACCATGGGCA AATATTATAC GCAAGGCGAC AAGGTGCTGA 900
TGCCGCTGGC GATTCAGGTT CATCATGCCG TCTGTGATGG CTTCCATGTC GGCAGAATGC 960
TTAATGAATT ACAACAGTAC TGCGATGAGT GGCAGGGCGG GGCGTAATTT TTTTAAGGCA 1020
GTTATTGGTG CCCTTAAAAC GCCTGGTGGA TCCTCTAGAG TCGACCTGCA GGCATGCAAG 1080
CTTGGCTGTT TTGGCGGATG AGAGAAGATT TTCAGCCTGA TACAGATTAA ATCAGAACGC 1140
AGAAGCGGTC TGATAAAACA GAATTTGCCT GGCGGCAGTA GCGCGGTGGT CCCACCTGAC 1200
CCCATGCCGA ACTCAGAAGT GAAACGCCGT AGCGCCGATG GTAGTGTGGG GTCTCCCCAT 1260
GCGAGAGTAG GGAACTGCCA GGCATCAAAT AAAACGAAAG GCTCAGTCGA AAGACTGGGC 1320
CTTTCGTTTT ATCTGTTGTT TGTCGGTGAA CGCTCTCCTG AGTAGGACAA ATCCGCCGGG 1380
AGCGGATTTG AACGTTGCGA AGCAACGGCC CGGAGGGTGG CGGGCAGGAC GCCCGCCATA 1440
AACTGCCAGG CATCAAATTA AGCAGAAGGC CATCCTGACG GATGGCCTTT TTGCGTTTCT 1500
ACAAACTCTT TTTGTTTATT TTTCTAAATA CATTCAAATA TGTATCCGCT CATGAGACAA 1560
TAACCCTGAT AAATGCTTCA ATAATATTGA AAAAGGAAGA GTATGAGTAT TCAACATTTC 1620
CGTGTCGCCC TTATTCCCTT TTTTGCGGCA TTTTGCCTTC CTGTTTTTGC TCACCCAGAA 1680
ACGCTGGTGA AAGTAAAAGA TGCTGAAGAT CAGTTGGGTG CACGAGTGGG TTACATCGAA 1740
CTGGATCTCA ACAGCGGTAA GATCCTTGAG AGTTTTCGCC CCGAAGAACG TTTTCCAATG 1800
ATGAGCACTT TTAAAGTTCT GCTATGTGGC GCGGTATTAT CCCGTGTTGA CGCCGGGCAA 1860
GAGCAACTCG GTCGCCGCAT ACACTATTCT CAGAATGACT TGGTTGAGTA CTCACCAGTC 1920
ACAGAAAAGC ATCTTACGGA TGGCATGACA GTAAGAGAAT TATGCAGTGC TGCCATAACC 1980
ATGAGTGATA ACACTGCGGC CAACTTACTT CTGACAACGA TCGGAGGACC GAAGGAGCTA 2040
ACCGCTTTTT TGCACAACAT GGGGGATCAT GTAACTCGCC TTGATCGTTG GGAACCGGAG 2100
CTGAATGAAG CCATACCAAA CGACGAGCGT GACACCACGA TGCCTACAGC AATGGCAACA 2160
ACGTTGCGCA AACTATTAAC TGGCGAACTA CTTACTCTAG CTTCCCGGCA ACAATTAATA 2220
GACTGGATGG AGGCGGATAA AGTTGCAGGA CCACTTCTGC GCTCGGCCCT TCCGGCTGGC 2280
TGGTTTATTG CTGATAAATC TGGAGCCGGT GAGCGTGGGT CTCGCGGTAT CATTGCAGCA 2340
CTGGGGCCAG ATGGTAAGCC CTCCCGTATC GTAGTTATCT ACACGACGGG GAGTCAGGCA 2400
ACTATGGATG AACGAAATAG ACAGATCGCT GAGATAGGTG CCTCACTGAT TAAGCATTGG 2460
TAACTGTCAG ACCAAGTTTA CTCATATATA CTTTAGATTG ATTTAAAACT TCATTTTTAA 2520
TTTAAAAGGA TCTAGGTGAA GATCCTTTTT GATAATCTCA TGACCAAAAT CCCTTAACGT 2580
GAGTTTTCGT TCCACTGAGC GTCAGACCCC GTAGAAAAGA TCAAAGGATC TTCTTGAGAT 2640
CCTTTTTTTC TGCGCGTAAT CTGCTGCTTG CAAACAAAAA AACCACCGCT ACCAGCGGTG 2700
GTTTGTTTGC CGGATCAAGA GCTACCAACT CTTTTTCCGA AGGTAACTGG CTTCAGCAGA 2760
GCGCAGATAC CAAATACTGT CCTTCTAGTG TAGCCGTAGT TAGGCCACCA CTTCAAGAAC 2820
TCTGTAGCAC CGCCTACATA CCTCGCTCTG CTAATCCTGT TACCAGTGGC TGCTGCCAGT 2880
GGCGATAAGT CGTGTCTTAC CGGGTTGGAC TCAAGACGAT AGTTACCGGA TAAGGCGCAG 2940
CGGTCGGGCT GAACGGGGGG TTCGTGCACA CAGCCCAGCT TGGAGCGAAC GACCTACACC 3000
GAACTGAGAT ACCTACAGCG TGAGCTATGA GAAAGCGCCA CGCTTCCCGA AGGGAGAAAG 3060
GCGGACAGGT ATCCGGTAAG CGGCAGGGTC GGAACAGGAG AGCGCACGAG GGAGCTTCCA 3120
GGGGGAAACG CCTGGTATCT TTATAGTCCT GTCGGGTTTC GCCACCTCTG ACTTGAGCGT 3180
CGATTTTTGT GATGCTCGTC AGGGGGGCGG AGCCTATGGA AAAACGCCAG CAACGCGGCC 3240
TTTTTACGGT TCCTGGCCTT TTGCTGGCCT TTTGCTCACA TGTTCTTTCC TGCGTTATCC 3300
CCTGATTCTG TGGATAACCG TATTACCGCC TTTGAGTGAG CTGATACCGC TCGCCGCAGC 3360
CGAACGACCG AGCGCAGCGA GTCAGTGAGC GAGGAAGCGG AAGAGCGCCT GATGCGGTAT 3420
TTTCTCCTTA CGCATCTGTG CGGTATTTCA CACCGCATAA TTTTGTTAAA ATTCGCGTTA 3480
AATTTTTGTT AAATCAGCTC ATTTTTTAAC CAATAGGCCG AAATCGGCAA AATCCCTTAT 3540
AAATCAAAAG AATAGACCGA GATAGGGTTG AGTGTTGTTC CAGTTTGGAA CAAGAGTCCA 3600
CTATTAAAGA ACGTGGACTC CAACGTCAAA GGGCGAAAAA CCGTCTATCA GGGCGATGGC 3660
CCACTACGTG AACCATCACC CTAATCAAGT TTTTTGGGGT CGAGGTGCCG TAAAGCACTA 3720
AATCGGAACC CTAAAGGGAG CCCCCGATTT AGAGCTTGAC GGGGAAAGCC GGCGAACGTG 3780
GCGAGAAAGG AAGGGAAGAA AGCGAAAGGA GCGGGCGCTA GGGCGCTGGC AAGTGTAGCG 3840
GTCACGCTGC GCGTAACCAC CACACCCGCC GCGCTTAATG CGCCGCTACA GGGCGCGTCC 3900
CATTCGCCAT TCAGGCTGCT ATGGTGCACT CTCAGTACAA TCTGCTCTGA TGCCGCATAG 3960
TTAAGCCAGT ATACACTCCG CTATCGCTAC GTGACTGGGT CATGGCTGCG CCCCGACACC 4020
CGCCAACACC CGCTGACGCG CCCTGACGGG CTTGTCTGCT CCCGGCATCC GCTTACAGAC 4080
AAGCTGTGAC CGTCTCCGGG AGCTGCATGT GTCAGAGGTT TTCACCGTCA TCACCGAAAC 4140
GCGCGAGGCA GCAGATCAAT TCGCGCGCGA AGGCGAAGCG GCATGCATTT ACGTTGACAC 4200
CATCGAATGG TGCAAAACCT TTCGCGGTAT GGCATGATAG CGCCCGGAAG AGAGTCAATT 4260
CAGGGTGGTG AATGTGAAAC CAGTAACGTT ATACGATGTC GCAGAGTATG CCGGTGTCTC 4320
TTATCAGACC GTTTCCCGCG TGGTGAACCA GGCCAGCCAC GTTTCTGCGA AAACGCGGGA 4380
AAAAGTGGAA GCGGCGATGG CGGAGCTGAA TTACATTCCC AACCGCGTGG CACAACAACT 4440
GGCGGGCAAA CAGTCGTTGC TGATTGGCGT TGCCACCTCC AGTCTGGCCC TGCACGCGCC 4500
GTCGCAAATT GTCGCGGCGA TTAAATCTCG CGCCGATCAA CTGGGTGCCA GCGTGGTGGT 4560
GTCGATGGTA GAACGAAGCG GCGTCGAAGC CTGTAAAGCG GCGGTGCACA ATCTTCTCGC 4620
GCAACGCGTC AGTGGGCTGA TCATTAACTA TCCGCTGGAT GACCAGGATG CCATTGCTGT 4680
GGAAGCTGCC TGCACTAATG TTCCGGCGTT ATTTCTTGAT GTCTCTGACC AGACACCCAT 4740
CAACAGTATT ATTTTCTCCC ATGAAGACGG TACGCGACTG GGCGTGGAGC ATCTGGTCGC 4800
ATTGGGTCAC CAGCAAATCG CGCTGTTAGC GGGCCCATTA AGTTCTGTCT CGGCGCGTCT 4860
GCGTCTGGCT GGCTGGCATA AATATCTCAC TCGCAATCAA ATTCAGCCGA TAGCGGAACG 4920
GGAAGGCGAC TGGAGTGCCA TGTCCGGTTT TCAACAAACC ATGCAAATGC TGAATGAGGG 4980
CATCGTTCCC ACTGCGATGC TGGTTGCCAA CGATCAGATG GCGCTGGGCG CAATGCGCGC 5040
CATTACCGAG TCCGGGCTGC GCGTTGGTGC GGATATCTCG GTAGTGGGAT ACGACGATAC 5100
CGAAGACAGC TCATGTTATA TCCCGCCGTT AACCACCATC AAACAGGATT TTCGCCTGCT 5160
GGGGCAAACC AGCGTGGACC GCTTGCTGCA ACTCTCTCAG GGCCAGGCGG TGAAGGGCAA 5220
TCAGCTGTTG CCCGTCTCAC TGGTGAAAAG AAAAACCACC CTGGCACCCA ATACGCAAAC 5280
CGCCTCTCCC CGCGCGTTGG CCGATTCATT AATGCAGCTG GCACGACAGG TTTCCCGACT 5340
GGAAAGCGGG CAGTGAGCGC AACGCAATTA ATGTGAGTTA GCGCGAATTG ATCTG 5395
|